Skip to Content
Merck

4.80864

SAFC

Potassium hydroxide solution 46-48%

EMPROVE® EXPERT

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing

Select a Size

25 L
€906.00
512 L
Inquire

€906.00


Estimated to ship on25 May 2025


Request a Bulk OrderRequest a Sample

Select a Size

Change View
25 L
€906.00
512 L
Inquire

About This Item

UNSPSC Code:
12352305

€906.00


Estimated to ship on25 May 2025


Request a Bulk OrderRequest a Sample

Quality Level

product line

EMPROVE® EXPERT

form

liquid

pH

>13.5 (20 °C in H2O)

application(s)

parenterals
pharmaceutical

storage temp.

2-30°C

General description

We offer a range of high quality critical raw materials, solvents and reagents plus the documentation and expertise to meet stringent regulatory requirements. In addition to our comprehensive portfolio, we also offer custom manufacturing services to meet your exact needs. As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Potassium hydroxide solution 46-48% is used for pH adjustment and for cleaning in place. Potassium hydroxide solution 46-48% Emprove® Expert is suitable for pharmaceutical and biopharmaceutical applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service