Skip to Content
Merck

1.06187

SAFC

Meglumine

EMPROVE® EXPERT, Ph. Eur., ChP, JP, USP

Pharma Manufacturing

Synonym(s):

N-Methyl-D-glucamine, 1-Deoxy-1-methylaminosorbitol, 1-Deoxy-1-methylaminosorbitol, 1-Deoxy-1-(methylamino)-D-glucitol, Meglumine

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€210.00
25 KG
€1,866.00

€210.00


Estimated to ship on02 April 2025


Request a Bulk OrderRequest a Sample

Select a Size

Change View
1 KG
€210.00
25 KG
€1,866.00

About This Item

Empirical Formula (Hill Notation):
C7H17NO5
CAS Number:
Molecular Weight:
195.21
Beilstein:
385906
MDL number:
UNSPSC Code:
12352104
EC Index Number:
228-506-9

€210.00


Estimated to ship on02 April 2025


Request a Bulk OrderRequest a Sample

Agency

ChP
JP
Ph. Eur.
USP

Quality Level

product line

EMPROVE® EXPERT

form

solid

autoignition temp.

~662 °F

pH

11 (20 °C, 10 g/L in H2O)

bp

210 °C/1013 hPa

mp

129-131.5 °C (lit.)

application(s)

liquid formulation
pharmaceutical
solid formulation
solubility enhancement

storage temp.

no temp limit

SMILES string

CNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C7H17NO5/c1-8-2-4(10)6(12)7(13)5(11)3-9/h4-13H,2-3H2,1H3/t4-,5+,6+,7+/m0/s1

InChI key

MBBZMMPHUWSWHV-BDVNFPICSA-N

Looking for similar products? Visit Product Comparison Guide

General description

When working on a pharmaceutical formulation, there are specific considerations to make and challenging hurdles to take before you can successfully launch your final drug product - formulation stability, release kinetics and bioavailability limitations just to name a few. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Meglumine Emprove® Essential is a robust pharmaceutical excipient that can provide stability, solubility enhancement and pH modification in solid and liquid formulations.

Features and Benefits

  • Solubility and bioavailability enhancement of APIs
  • Excellent counterion performance, a viable alternative to sodium
  • API grade: manufactured under the cGMP ICH Q7 guideline for APIs
  • Sole manufacturing location for API-grade meglumine in Europe
  • Multi-compendial product, complying with all major pharmacopeias, including ChP
  • Meglumine Emprove® API has also been successfully co-reviewed in China

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This overview describes the challenges when developing parenteral formulations and strategies to overcome them and reduce the risk of production.

Our overview of API solubility and dissolution enhancement methods can help you decide between the many options to take during formulation.

This article describes the use of excipients to help address API stability issues.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service