Skip to Content
Merck
Meglumine European Pharmacopoeia (EP) Reference Standard

Meglumine

Meglumine European Pharmacopoeia (EP) Reference Standard
Synonym(s):
N-Methyl-D-glucamine, 1-Deoxy-1-(methylamino)-D-glucitol, Meglumine
Empirical Formula (Hill Notation):
C7H17NO5
CAS Number:
Molecular Weight:
195.21

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
API Family
Reference Material Type
Special Grade
Technique