Skip to Content
Merck

1.11597

SAFC

Parteck® SI 400 LEX (Sorbitol)

EMPROVE® EXPERT, Ph. Eur., BP, NF, JP

Pharma Manufacturing

Synonym(s):

Parteck® Sorbitol, D-Glucitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
MDL number:
UNSPSC Code:
12352201
EC Index Number:
200-061-5
Pricing and availability is not currently available.

Agency

BP
JP
NF
Ph. Eur.

Quality Level

product line

EMPROVE® EXPERT

form

solid

autoignition temp.

420 °C

pH

6-7 (20 °C, 100 g/L in H2O)

mp

94-96 °C

transition temp

flash point >100 °C

solubility

2350 g/L

density

1.49 g/cm3 at 20 °C

application(s)

liquid formulation
ophthalmics
parenterals
pharmaceutical

storage temp.

2-25°C

SMILES string

OC(C(O)C(O)CO)C(O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2

InChI key

FBPFZTCFMRRESA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Sorbitol is a sugar alcohol which is widely used in biomolecule formulations as a stabilizer and tonicity agent.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
PARTECK is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This overview describes the challenges when developing parenteral formulations and strategies to overcome them and reduce the risk of production.

This article outlines critical factors in long-term stability of proteins, and how different stabilizers protect again mechanical and thermal stress.

Related Content

Learn more on mAb subcutaneous or intravenous formulation, and the relevant associated products, including excipients.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service