Skip to Content
Merck
All Photos(1)

Key Documents

1.05051

Supelco

Potassium iodate

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Potassium iodate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€66.50
500 G
€144.00

€66.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€66.50
500 G
€144.00

About This Item

Empirical Formula (Hill Notation):
IKO3
CAS Number:
Molecular Weight:
214.00
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-831-9
NACRES:
NA.21
grade:
ACS reagent
form:
solid
solubility:
92 g/L

€66.50


Check Cart for Availability

Request a Bulk Order

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

product line

EMSURE®

Assay

99.7-100.4% (iodometric)

form

solid

impurities

≤0.005% Insoluble matter

loss

≤0.05% loss on drying, 130 °C

pH

6 (20 °C, 50 g/L in H2O)

mp

560 °C (decomposes)

solubility

92 g/L

density

3.98 g/cm3 at 20 °C

bulk density

2000 kg/m3

anion traces

bromide, bromate, chloride, chlorate (as Cl-): ≤0.01%
iodide (I-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

As: ≤0.0003%
Fe: ≤0.001%
N: ≤0.002%
Na: ≤0.005%
Pb: ≤0.0004%
heavy metals: ≤5 ppm
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

[K+].[I](=O)(=O)[O-]

InChI

1S/HIO3.K/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

JLKDVMWYMMLWTI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Evaluation of the chemical composition and nutritional value of lettuce biofortified with iodine: This research examines the impact of potassium iodate as an iodine source for hydroponically grown lettuce, enhancing its nutritional value which is significant for dietary iodine intake (Dyląg A et al., 2023).

  • Selected adjuvants increase the efficacy of foliar biofortification of iodine in bread wheat: Potassium iodate used in foliar sprays significantly improves iodine content in wheat, demonstrating its potential to enhance the nutritional quality of basic food staples (Magor E et al., 2023).

  • Quality of Rye Plants as Affected by Agronomic Biofortification with Iodine: Investigates how potassium iodate affects the growth and iodine content of rye plants, providing insights into iodine biofortification practices in agriculture (Krzepiłko A et al., 2022).

  • Anti- and pro-oxidant potential of lettuce biofortified with iodine: Studies the antioxidant properties of lettuce treated with potassium iodate, contributing valuable data for food science, particularly in enhancing the health benefits of vegetables (Sularz O et al., 2021).

  • Effectiveness of enriching lettuce with iodine using salicylic and diiodosalicylic acids and the chemical composition of plants: Highlights the use of potassium iodate in modifying the nutrient profile of lettuce, aiming at boosting iodine levels through agricultural practices (Smoleń S et al., 2022).

Analysis Note

Assay (iodometric): 99.7 - 100.4 %
Insoluble matter: ≤ 0.005 %
pH-value (5 %; water): 5.0 - 8.0
Chloride, Chlorate, Bromide, Bromate (as Cl): ≤ 0.01 %
Iodide (I): ≤ 0.001 %
Sulfate (SO₄): ≤ 0.005 %
Total nitrogen (N): ≤ 0.002 %
Heavy metals (ACS): ≤ 5 ppm
Heavy metals (as Pb): ≤ 0.0005 %
As (Arsenic): ≤ 0.0003 %
Fe (Iron): ≤ 0.001 %
Na (Sodium): ≤ 0.005 %
Pb (Lead): ≤ 0.0004 %
Loss on drying (130 °C): ≤ 0.05 %
Corresponds to ACS, ISO, Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service