Skip to Content
Merck
All Photos(2)

Documents

1.06392

Supelco

Sodium carbonate

anhydrous for analysis EMSURE® ISO

Synonym(s):

anhydrous soda, Calcined soda, Carbonic acid disodium salt, Soda ash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2CO3
CAS Number:
Molecular Weight:
105.99
Beilstein:
4154566
MDL number:
UNSPSC Code:
12352302
EC Index Number:
207-838-8
NACRES:
NA.21

Agency

reag. ISO
suitable for SM 2310
suitable for SM 2320
suitable for SM 4500 - NH3

Quality Level

product line

EMSURE®

Assay

≥99.9% dry basis (acidimetric)

form

powder

potency

4090 mg/kg LD50, oral (Rat)

loss

≤1% loss on drying, 300 °C

pH

11.16 (25 °C, 4 g/L in H2O)

mp

851 °C (lit.)

solubility

212.5 g/L

density

2.53 g/cm3 at 20 °C

bulk density

1100 kg/m3

anion traces

chloride (Cl-): ≤0.002%
phosphate (PO43-): ≤0.001%
silicate (as SiO2): ≤0.002%
sulfur compounds (as SO42-): ≤0.005%

cation traces

Al: ≤0.001%
Ca: ≤0.005%
Fe: ≤0.0005%
K: ≤0.01%
Mg: ≤0.0005%
N: ≤0.001%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

O=C(O[Na])O[Na]

InChI

1S/CH2O3.2Na/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

CDBYLPFSWZWCQE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Enhancing Carbonate Content in Hydroxyapatites: Sodium carbonate is crucial in synthesizing carbonate-substituted hydroxyapatites, which are important materials in bone regeneration and dental applications. Recent research has focused on maximizing the carbonate content in these materials, highlighting the role of sodium carbonate in enhancing their structural and functional properties (Nowicki et al., Journal of Solid State Chemistry, 2021).

Biochem/physiol Actions

Buffer component, may be used for the removal of peripheral membrane proteins.

Analysis Note

Assay (acidimetric; calculated on dried substance): ≥ 99.9 %
Chloride (Cl): ≤ 0.002 %
Phosphate (PO₄): ≤ 0.001 %
Silicate (as SiO₂): ≤ 0.002 %
Total sulfur (as SO₄): ≤ 0.005 %
Total nitrogen (N): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.0005 %
Al (Aluminium): ≤ 0.001 %
Ca (Calcium): ≤ 0.005 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.01 %
Mg (Magnesium): ≤ 0.0005 %
Loss on drying (300 °C): ≤ 1.0 %
Corresponds to ISO

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service