Skip to Content
Merck
All Photos(1)

Key Documents

1.09914

Supelco

Iodide iodate solution

c(I₂) = 1/128 mol/l (1/64 N), concentrate for preparing 1000 mL volumetric solution for titration, Titrisol®

Sign Into View Organizational & Contract Pricing

Select a Size

1 AMP
€41.40

€41.40


Estimated to ship on21 May 2025


Request more information

Select a Size

Change View
1 AMP
€41.40

About This Item

UNSPSC Code:
12352402
NACRES:
NA.21

€41.40


Estimated to ship on21 May 2025


Request more information

Product Name

Iodide iodate solution, for 1000 ml, c(I₂) = 1/128 mol/l (1/64 N) Titrisol®

Quality Level

product line

Titrisol®

form

liquid

reaction suitability

reaction type: Redox Reactions

concentration

1/128 M

technique(s)

titration: suitable

pH

13 (20 °C in H2O)

density

1.07 g/cm3 at 20 °C

storage temp.

15-25°C

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT RE 2 Oral

Target Organs

Thyroid

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service