Skip to Content
Merck
All Photos(1)

Key Documents

1.05050

SAFC

Potassium iodate

extra pure FCC

Synonym(s):

Potassium iodate

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€300.00
25 KG
€8,000.00

€300.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
500 G
€300.00
25 KG
€8,000.00

About This Item

Empirical Formula (Hill Notation):
IKO3
CAS Number:
Molecular Weight:
214.00
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-831-9

€300.00


Estimated to ship on25 May 2025


Request a Bulk Order

reg. compliance

FCC

Quality Level

form

crystalline

pH

6 (20 °C, 50 g/L in H2O)

mp

560 °C (decomposes)

solubility

92 g/L

density

3.98 g/cm3 at 20 °C

bulk density

2000 kg/m3

application(s)

food and beverages

storage temp.

15-25°C

SMILES string

[K+].[I](=O)(=O)[O-]

InChI

1S/HIO3.K/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

JLKDVMWYMMLWTI-UHFFFAOYSA-M

Related Categories

General description

We offer a range of high quality critical raw materials plus the documentation and expertise to help our customers meet stringent regulatory requirements. Our SAFC® portfolio of high-quality products withstands strict quality control procedures.

Application

Potassium iodate extra pure is used as a reagent and as food additive.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service