Skip to Content
Merck
All Photos(1)

Key Documents

1.04867

Supelco

Potassium hydrogen diiodate

for analysis EMSURE®

Synonym(s):

Potassium hydrogen diiodate, Potassium biiodate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KH(IO3)2
CAS Number:
Molecular Weight:
389.91
MDL number:
UNSPSC Code:
12352302
EC Index Number:
236-650-9
Assay:
≥99.8% (iodometric)
form:
solid
solubility:
13 g/L

Quality Level

product line

EMSURE®

Assay

≥99.8% (iodometric)

form

solid

pH

1-2 (20 °C, 50 g/L in H2O)

solubility

13 g/L

bulk density

1750 kg/m3

anion traces

bromide, bromate, chloride, chlorate (as Cl-): ≤0.02%

cation traces

Fe: ≤0.0005%
N: ≤0.002%
heavy metals (as Pb): ≤0.001%

storage temp.

2-30°C

InChI

1S/2HIO3.K/c2*2-1(3)4;/h2*(H,2,3,4);/q;;+1/p-1

InChI key

ACAYDTMSDROWHW-UHFFFAOYSA-M

Analysis Note

Assay (iodometric): ≥ 99.8 %
Chloride, Chlorate, Bromide, Bromate (as Cl): ≤ 0.02 %
Total nitrogen (N): ≤ 0.002 %
Heavy metals (as Pb): ≤ 0.001 %
Fe (Iron): ≤ 0.0005 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 2 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service