Skip to Content
Merck
All Photos(1)

Key Documents

1.06541

Supelco

Sodium nitroprusside dihydrate

[disodium pentacyanonitrosyl ferrate(III) dihydrate] GR for analysis ACS,Reag. Ph Eur

Synonym(s):

Sodium nitroprusside dihydrate, Nitroprusside sodium, Sodium nitroferricyanide

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€71.30
100 G
€144.00
500 G
€567.00

€71.30


Estimated to ship on28 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
25 G
€71.30
100 G
€144.00
500 G
€567.00

About This Item

Linear Formula:
Na2[Fe(CN)5NO] · 2H2O
CAS Number:
Molecular Weight:
281.95
MDL number:
UNSPSC Code:
12352302
EC Index Number:
238-373-9
NACRES:
SE.11
grade:
ACS reagent
solubility:
400 g/L (slow decomposition)

€71.30


Estimated to ship on28 May 2025


Request a Bulk OrderRequest more information

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

potency

99 mg/kg LD50, oral (Rat)

specific analyte(s)

ammonium

technique(s)

photometry: suitable

pH

5 (20 °C, 50 g/L in H2O)

solubility

400 g/L (slow decomposition)

density

1.71 g/cm3 at 20 °C

bulk density

1000 kg/m3

storage temp.

2-30°C

InChI

1S/5CN.Fe.NO.2Na.2H2O/c5*1-2;;1-2;;;;/h;;;;;;;;;2*1H2/q;;;;;-2;;2*+1;;

InChI key

NWFKTGARQFKZMN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • SPECTROPHOTOMETRIC DETERMINATION OF THYMOL IN MOUTHWASHES VIA SODIUM NITROPRUSSIDE REACTION WITH HYDROXIDE AMINE HYDROCHLORIDE.: This study presents a spectrophotometric method for determining thymol in mouthwashes using a reaction with sodium nitroprusside and hydroxide amine hydrochloride. This analytical approach enhances the accuracy and reliability of thymol quantification in pharmaceutical products (International Journal of Research in Pharmacy and Chemistry, 2023).

  • Efficacy of postharvest sodium nitroprusside application to extend storability by regulating physico-chemical quality of pear fruit.: This study investigates the application of sodium nitroprusside in extending the storability of pear fruits by regulating their physico-chemical quality. The findings suggest significant improvements in fruit preservation and quality maintenance during storage (Food Chemistry, 2021).

Linkage

Replaces: SX0675-1; SX0675-2; SX0675

Analysis Note

Assay (argentometric): 99.0 - 102.0 %
Identity: passes test
Insoluble matter: ≤ 0.01 %
Chloride (Cl): ≤ 0.02 %
Hexacyanoferrate (II): ≤ 0.02 %
Hexacyanoferrate (III): ≤ 0.01 %
Sulfate (SO₄): ≤ 0.01 %
Corresponds to ACS, Reag. Ph. Eur.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–3 of 3 Questions  
  1. Can I use it to prepare a solution to inject via i.p. in rodents?

    1 answer
    1. This product is intended for research use only and is not suitable for human or veterinary use. While not tested for in vitro or in vivo use, this compound has been used successfully in cell culture and animal models by other customers. Please see the links below to review example publications:
      Rat animal model - https://pmc.ncbi.nlm.nih.gov/articles/PMC3556448/
      Rat hepatocytes - https://pmc.ncbi.nlm.nih.gov/articles/PMC117951/
      Porcine lung & mesentery - https://pmc.ncbi.nlm.nih.gov/articles/PMC1572630/

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service