Skip to Content
Merck
All Photos(2)

Key Documents

1.05033

Supelco

Potassium hydroxide

pellets for analysis EMSURE®

Synonym(s):

Potash caustic, Caustic potash

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€49.20
1 KG
€89.70
5 KG
€335.00
25 KG
€835.00
50 KG
€1,670.00

€49.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 G
€49.20
1 KG
€89.70
5 KG
€335.00
25 KG
€835.00
50 KG
€1,670.00

About This Item

Linear Formula:
KOH
CAS Number:
Molecular Weight:
56.11
MDL number:
UNSPSC Code:
12352305
EC Index Number:
215-181-3
NACRES:
NA.21
form:
solid
solubility:
1130 g/L

€49.20


Check Cart for Availability

Request a Bulk Order

Agency

suitable for SM 5210

Quality Level

vapor pressure

1 mmHg ( 719 °C)

product line

EMSURE®

Assay

≥85.0% KOH basis (acidimetric)

form

solid

potency

333 mg/kg LD50, oral (Rat)

pH

14 (20 °C, 56 g/L in H2O)

mp

361 °C (lit.)

solubility

1130 g/L

anion traces

carbonate (K2CO3): ≤1.0%
chloride (Cl-): ≤0.005%
phosphate (PO43-): ≤0.0005%
silicate (as SiO2): ≤0.005%
sulfate (SO42-): ≤0.0005%

cation traces

Al: ≤0.001%
Ca: ≤0.001%
Cu: ≤0.0002%
Fe: ≤0.0005%
N: ≤0.0005%
Na: ≤0.5%
Ni: ≤0.0005%
Pb: ≤0.0005%
Zn: ≤0.0025%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

[OH-].[K+]

InChI

1S/K.H2O/h;1H2/q+1;/p-1

InChI key

KWYUFKZDYYNOTN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (acidimetric, KOH): ≥ 85.0 %
Carbonate (as K₂CO₃): ≤ 1.0 %
Chloride (Cl): ≤ 0.005 %
Phosphate (PO₄): ≤ 0.0005 %
Silicate (SiO₂): ≤ 0.005 %
Sulfate (SO₄): ≤ 0.0005 %
Total nitrogen (N): ≤ 0.0005 %
Heavy metals (as Pb): ≤ 0.0005 %
Al (Aluminium): ≤ 0.001 %
Ca (Calcium): ≤ 0.001 %
Cu (Copper): ≤ 0.0002 %
Fe (Iron): ≤ 0.0005 %
Na (Sodium): ≤ 0.5 %
Ni (Nickel): ≤ 0.0005 %
Pb (Lead): ≤ 0.0005 %
Zn (Zinc): ≤ 0.0025 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Compare PAH determination in olive oil: traditional GB 5009.265-2021 method vs. faster, simpler SPE with Supelclean EZ-POP NP; both meet criteria.

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service