Skip to Content
Merck

Skip To

07-722

Anti-phospho-VEGFR2/Flk-1/KDR (Tyr1054) Antibody

Upstate®, from rabbit

Synonym(s):

Kinase insert domain receptor, Protein-tyrosine kinase receptor Flk-1, kinase insert domain receptor (a type III receptor tyrosine kinase), vascular endothelial growth factor receptor 2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 357,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
polyclonal
Species reactivity:
zebrafish, human
Application:
WB
Citations:
3

€ 357,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

zebrafish, human

species reactivity (predicted by homology)

primate (based on 100% sequence homology), horse (based on 100% sequence homology), rat (based on 100% sequence homology), mouse (based on 100% sequence homology), chicken (based on 100% sequence homology), bovine (based on 100% sequence homology), Danio (based on 100% sequence homology)

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pTyr1054)

Gene Information

human ... KDR(3791)

General description

152 kDa
The Flt gene family of receptor tyrosine kinases includes Flt-1, KDR/Flk-1, Flt-3, and Flt-4. They encode receptors for VEGF, characterized by seven Ig-loops within their extracellular domain and a split kinase domain within the cytoplasmic domain. VEGF is an important angiogenic factor of many solid tumors and is also involved in the differentiation of endothelial cells. Flt-1 (Fms-like tyrosine kinase, VEGF-R1) is expressed on endothelial cells and on the common precursors of endothelial and hematopoietic stem cells. KDR (Flk-1, VEGF-R2) is the principal receptor involved in VEGF signaling. KDR is expressed on endothelial cells and stimulates angiogenesis, but is also expressed on bone marrow cells, and is thought to play a role in driving the proliferation of most or all leukemic cells. Flt3 (also known as Flk-2) is a class III receptor tyrosine kinase that is closely related to c-Kit and c-Fms. It is expressed in primitive hematopoietic cells and its ligand exerts proliferative effects on these cells in co-operation with other cytokines.

Immunogen

Synthetic peptide encompassing

Application

Anti-phospho-VEGFR2/Flk-1/KDR (Tyr1054) Antibody is an antibody against phospho-VEGFR2/Flk-1/KDR (Tyr1054) for use in WB.
Research Category
Signaling

Biochem/physiol Actions

Recognizes VEGFR2/Flk-1/KDR when phosphorylated on Tyrosine 1054 (human).

Physical form

Affinity purified rabbit polyclonal IgG in PBS with 0.05% NaN3 and 50% glycerol.
Double affinity purified

Preparation Note

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
CSF-1 treated PAE cell lysate, transfected with CSF/KDR.
Western Blot Analysis:
This lot detected VEGFR2 phosphorylated on Tyr1054 at a 1:1,000 dilution in PAE cells transfected with CSF/KDR and treated with 40ng/mL CSF-1 for 15 min. Cell lysate resolved via SDS-PAGE and transferred to PVDF (Immobilon-P).

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
07-716-IABS55307-758
antibody form

affinity purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

species reactivity

zebrafish, human

species reactivity

mouse, human

species reactivity

human, mouse

species reactivity

bovine, human, goat, sheep, rat, mouse

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

Gene Information

human ... KDR(3791)

Gene Information

human ... KDR(3791)
mouse ... Kdr(16542)

Gene Information

human ... KDR(3791)

Gene Information

bovine ... Flt1(503620)
human ... FLT1(2321)
mouse ... Flt1(14254)
rat ... Flt1(54251)

UniProt accession no.

P35968

UniProt accession no.

P35918

UniProt accession no.

P35968

UniProt accession no.

P17948


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
07-72204053252403668

Questions

Reviews

No rating value

Active Filters