Skip to Content
Merck
All Photos(2)

Key Documents

07-716-I

Sigma-Aldrich

Anti-KDR/Flk-1/VEGFR2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Vascular endothelial growth factor receptor 2, VEGFR-2, Fetal liver kinase 1, FLK-1, Kinase NYK, Protein-tyrosine kinase receptor flk-1, CD309

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€421.00

€421.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€421.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€421.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KDR(3791)
mouse ... Kdr(16542)

General description

The Flt gene family of receptor tyrosine kinases includes Flt-1, KDR/Flk-1, Flt-3, and Flt-4. They encode receptors for VEGF, characterized by seven Ig-loops within their extracellular domain and a split kinase domain within the cytoplasmic domain. VEGF is an important angiogenic factor of many solid tumors and is also involved in the differentiation of endothelial cells. Flt-1 (Fms-like tyrosine kinase, VEGF-R1) is expressed on endothelial cells and on the common precursors of endothelial and hematopoietic stem cells. KDR (Flk-1, VEGF-R2) is the principal receptor involved in VEGF signaling. KDR is expressed on endothelial cells and stimulates angiogenesis, but is also expressed on bone marrow cells, and is thought to play a role in driving the proliferation of most or all leukemic cells. Flt3 is a class III receptor tyrosine kinase that is closely related to c-Kit and c-Fms. It is expressed in primitive hematopoietic cells and its ligand exerts proliferative effects on these cells in co-operation with other cytokines. The VEGF receptors stimulate the Ras-MAP kinase pathway, suggesting that it signals as a conventional receptor tyrosine kinase.

Immunogen

Epitope: C-terminus
GST-tagged recombinant protein corresponding to the C-terminus of mouse KDR/Flk-1/VEGFR2.

Application

Detect KDR/Flk-1/VEGFR2 using this rabbit polyclonal antibody, Anti-KDR/Flk-1/VEGFR2 Antibody validated for use in western blotting.
Research Category
Signaling
Research Sub Category
Developmental Signaling
Western Blotting Analysis: A 1:500 dilution from a representative lot detected KDR/Flk-1/VEGFR2 in 10 µg of PAE cells transfected with CSF/VEGFR2 chimera.

Quality

Evaluated by Western Blotting in HUVEC cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected KDR/Flk-1/VEGFR2 in 10 µg of HUVEC cell lysate.

Target description

~230 kDa observed

Linkage

Replaces: 07-716

Physical form

Affinity purified
Purified rabbit Polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edward Hartsough et al.
iScience, 25(8), 104736-104736 (2022-08-10)
Through tightly controlled multilayer mechanisms, vascular endothelial growth factor receptor-2 (VEGFR-2) activation and its downstream signal transduction govern vasculogenesis and pathological angiogenesis, such as tumor angiogenesis. Therefore, it is critical to understand the molecular mechanisms governing VEGFR-2 signal transduction. We
Chike Osude et al.
Cells, 11(10) (2022-05-29)
NSCLC treatment includes targeting of EGFR with tyrosine kinase inhibitors (TKIs) such as Erlotinib; however, resistance to TKIs is commonly acquired through T790M EGFR mutations or overexpression of vascular endothelial growth factor receptor-2 (VEGFR-2). We investigated the mechanisms of EGFR-TKI

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service