Skip to Content
Merck
All Photos(1)

Key Documents

860064P

Avanti

NGcGM3

Avanti Research - A Croda Brand 860064P, powder

Synonym(s):

NGcGM3 Ganglioside (bovine)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€288.00
500 μG
€878.00

€288.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€288.00
500 μG
€878.00

About This Item

Empirical Formula (Hill Notation):
C65H121N3O22
CAS Number:
Molecular Weight:
1296.66
UNSPSC Code:
12352211
NACRES:
NA.25

€288.00


Check Cart for Availability

Request a Bulk Order

Assay

≥99% (TLC)

form

powder

packaging

pkg of 1 × 100 μg (860064P-100UG)
pkg of 1 × 500 μg (860064P-500UG)

manufacturer/tradename

Avanti Research - A Croda Brand 860064P

lipid type

lipid extracts

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCC/C=C\CCCCCCCC)=O)CO[C@H](O1)[C@H](O)[C@@H](O)[C@H](O[C@@H]2O[C@H](CO)[C@@H]([C@H](O[C@@]3(C([O-])=O)O[C@@H]([C@H](O)[C@H](O)CO)[C@@](NC(CO)=O)([H])[C@@H](O)C3)[C@H]2O)O)[C@H]1CO.[NH4+]

InChI key

REEGKSVAHNARFK-XHKMRSCRSA-N

Packaging

5 mL Amber Glass Screw Cap Vial (860064P-100UG)
5 mL Amber Glass Screw Cap Vial (860064P-500UG)

Other Notes

Structure, molecular formula, and molecular weight are for a representative form of the material.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service