Skip to Content
Merck
All Photos(1)

Key Documents

8.41271

Sigma-Aldrich

2,3,5-Trichlorobenzaldehyde

for synthesis

Synonym(s):

2,3,5-Trichlorobenzaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€83.90

€83.90


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
€83.90

About This Item

Empirical Formula (Hill Notation):
C7H3Cl3O
CAS Number:
Molecular Weight:
209.46
MDL number:
UNSPSC Code:
12352114
NACRES:
NA.22

€83.90


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥97.0% (GC)

form

powder

mp

75-76 °C

storage temp.

2-30°C

SMILES string

Clc1c(cc(cc1C=O)Cl)Cl

InChI

1S/C7H3Cl3O/c8-5-1-4(3-11)7(10)6(9)2-5/h1-3H

InChI key

DJYRZTCLVDKWBL-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 73 °C
Melting range (upper value): ≤ 76 °C
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service