Skip to Content
Merck
All Photos(2)

Key Documents

840863C

Avanti

18:0-20:4 PA

1-stearoyl-2-arachidonoyl-sn-glycero-3-phosphate (sodium salt), chloroform

Synonym(s):

1-octadecanoyl-2--(5Z,8Z,11Z,14Z-eicosatetraenoyl)-sn-glycero-3-phosphate (sodium salt); SAPA; PA(18:0/20:4(5Z,8Z,11Z,14Z)); 110618

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€632.00
100 MG
€1,560.00

€632.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€632.00
100 MG
€1,560.00

About This Item

Empirical Formula (Hill Notation):
C41H72O8PNa
CAS Number:
Molecular Weight:
746.97
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

€632.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840863C-25mg)
pkg of 1 × 4 mL (840863C-100mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840863C

concentration

10 mg/mL (840863C-25mg)
25 mg/mL (840863C-100mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[P](=O)([O-])(OC[C@H](OC(=O)CCC\C=C/C\C=C/C\C=C/C\C=C/CCCCC)COC(=O)CCCCCCCCCCCCCCCCC)O

InChI

1S/C41H73O8P.Na/c1-3-5-7-9-11-13-15-17-19-20-22-24-26-28-30-32-34-36-41(43)49-39(38-48-50(44,45)46)37-47-40(42)35-33-31-29-27-25-23-21-18-16-14-12-10-8-6-4-2;/h11,13,17,19,22,24,28,30,39H,3-10,12,14-16,18,20-21,23,25-27,29,31-38H2,1-2H3,(H2,44,45,46);/q;+1/p-1/b13-11-,19-17-,24-22-,30-28-;/t39-;/m1./s1

InChI key

ZNLBWELULQXDSY-JNZHLZGHSA-M

General description

18:0-20:4 PA or 1-stearoyl-2-arachidonoyl-sn-glycero-3-phosphate (sodium salt) is a sodium salt of phosphatidic acid, with substitution of stearic acid and arachidonic acid at the sn-1 and sn-2 positions respectively, in the glycerol backbone.

Application

18:0-20:4 PA or 1-stearoyl-2-arachidonoyl-sn-glycero-3-phosphate (sodium salt) has been used in liposomes to determine the lipid-binding properties of the Praja-1 protein.[1]

Packaging

5 mL Clear Glass Sealed Ampule (840863C-100mg)
5 mL Clear Glass Sealed Ampule (840863C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiang Lu et al.
FEBS letters, 594(11), 1787-1796 (2020-03-07)
Serotonin transporter (SERT) is involved in serotonergic system regulation and in the pathophysiology/therapeutics of serotonin-/SERT-related diseases such as obsessive-compulsive disorder, depression, autism, and schizophrenia. We recently revealed that diacylglycerol (DG) kinase (DGK) δ induces ubiquitination/degradation of SERT in a DGK

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service