Skip to Content
Merck
All Photos(2)

Key Documents

855676C

Avanti

17:0 Lyso PC

1-heptadecanoyl-2-hydroxy-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1-heptadecanoyl-sn-glycero-3-phosphocholine; PC(17:0/0:0)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€181.00

€181.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€181.00

About This Item

Empirical Formula (Hill Notation):
C25H52NO7P
CAS Number:
Molecular Weight:
509.66
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

€181.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (855676C-25mg)
pkg of 2 × 4 mL (855676C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 855676C

concentration

10 mg/mL (855676C-25mg)
25 mg/mL (855676C-200mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OC[C@H](O)COC(=O)CCCCCCCCCCCCCCCC)OCC[N+](C)(C)C

InChI

1S/C25H52NO7P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-25(28)31-22-24(27)23-33-34(29,30)32-21-20-26(2,3)4/h24,27H,5-23H2,1-4H3/t24-/m1/s1

InChI key

SRRQPVVYXBTRQK-XMMPIXPASA-N

Application

17:0 Lyso PC has been used for the extraction of lipids from the blood samples.[1]

Packaging

5 mL Clear Glass Sealed Ampule (855676C-200mg)
5 mL Clear Glass Sealed Ampule (855676C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poorna C R Yalagala et al.
Molecular nutrition & food research, 64(12), e2000059-e2000059 (2020-04-19)
Currently available omega-3 fatty acid supplements do not enrich the docosahexaenoic acid (DHA) of the adult brain because they are absorbed as triacylglycerol, whereas the transporter at the blood brain barrier requires lysophosphatidylcholine (LPC)-DHA. The hypothesis that treatment of krill
Generation and quality control of lipidomics data for the alzheimer?s disease neuroimaging initiative cohort
Barupal DK, et al.
Scientific data, 5, 180263-180263 (2018)
Melania Gaggini et al.
International journal of molecular sciences, 20(24) (2020-01-01)
Metabolomics/lipidomics are important tools to identify novel biomarkers associated with liver damage. Patients with chronic liver disease (CLD) and hepatitis C virus (HCV) infection often have alterations in glucose, lipid and protein metabolism. The aim of this study was to
Hiroyasu Sato et al.
Cell reports, 31(5), 107579-107579 (2020-05-07)
Polyunsaturated fatty acids (PUFAs) confer health benefits by preventing inflammation and obesity and by increasing thermogenesis in brown and beige adipocytes. As well as being supplied exogenously as nutrients, PUFAs are largely stored in membrane glycerophospholipids and released by phospholipase
Michael M Dubreuil et al.
Cell reports, 30(5), 1417-1433 (2020-02-06)
Reactive oxygen species (ROS) play critical roles in metabolism and disease, yet a comprehensive analysis of the cellular response to oxidative stress is lacking. To systematically identify regulators of oxidative stress, we conducted genome-wide Cas9/CRISPR and shRNA screens. This revealed a detailed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service