Key Documents
Z680443
Duran® GL 45 connection system caps and accessories
hose connection screw cap, blue PP GL 14
Synonym(s):
1129814
Select a Size
2 EA
€9.20
€9.20
Check Cart for Availability
Select a Size
Change View
2 EA
€9.20
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
€9.20
Check Cart for Availability
material
blue polypropylene
manufacturer/tradename
DURAN® 1129814
parameter
-40-140 °C temp. limit
General description
GL 45 screw cap with either two or three ports that have GL 14 thread. This is a flexible modular system which has four different hose diameters available: 1.6 mm, 3.0 mm, 3.2 mm and 6.0 mm that can be connected.
Sterile pressure equalization is possible through use of membrane filter.
Features and Benefits
  • Manufactured from PP, PTFE, silicone
  • Accessories designed for use with the DURAN® GL 45 2/3 port tubing connection caps or GL 45 / GLS 80® Stirred Bottle Reactor caps
  • Safe and secure liquid transfer into or out of laboratory bottles
  • Four inserts are suitable for rigid or semi-rigid tubing with a range of outer diameters: 1.6, 3.0, 3.2 or 6.0 mm
  • Open-topped blue PP GL 14 pass-through tubing cap for use with the inserts
  • Closed-top red PBT GL 14 cap for sealing unused ports
  • Sterile pressure equilisation is possible with pressure compensation set
Legal Information
DURAN is a registered trademark of DWK Life Sciences
GLS 80 is a registered trademark of Duran Group
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. Does this item include all the pieces shown in the photo, or is that a different SKU?
    1 answer
    1. The photo provided is representative of the entire group of related GL 45 capping products. Each component is offered separately. This specific product is the blue hose connection screw cap shown second from the right as seen in provided image.
      Helpful?