Skip to Content
Merck
All Photos(1)

Key Documents

T14206

Sigma-Aldrich

cis-1,2,3,6-Tetrahydrophthalimide

96%

Synonym(s):

cis-4-Cyclohexene-1,2-dicarboximide

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€54.20

€54.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€54.20

About This Item

Empirical Formula (Hill Notation):
C8H9NO2
CAS Number:
Molecular Weight:
151.16
Beilstein:
82338
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€54.20


Check Cart for Availability

Request a Bulk Order

Assay

96%

mp

129-133 °C (lit.)

SMILES string

[H][C@]12CC=CC[C@@]1([H])C(=O)NC2=O

InChI

1S/C8H9NO2/c10-7-5-3-1-2-4-6(5)8(11)9-7/h1-2,5-6H,3-4H2,(H,9,10,11)/t5-,6+

InChI key

CIFFBTOJCKSRJY-OLQVQODUSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

352.4 °F - closed cup

Flash Point(C)

178.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service