Skip to Content
Merck
All Photos(3)

Key Documents

426288

Sigma-Aldrich

Tetrabutylammonium bromide

greener alternative

ACS reagent, ≥98.0%

Synonym(s):

N,N,N-tributyl-1-butanaminium bromide, TBAB, TBABr, tetra-n-butylammonium bromide

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€59.30
100 G
€84.00

€59.30


Estimated to ship on02 April 2025


Request a Bulk Order

Select a Size

Change View
25 G
€59.30
100 G
€84.00

About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(Br)
CAS Number:
Molecular Weight:
322.37
Beilstein:
3570983
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥98.0%
grade:
ACS reagent
form:
solid

€59.30


Estimated to ship on02 April 2025


Request a Bulk Order

grade

ACS reagent

Quality Level

Assay

≥98.0%

form

solid

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤0.5% tributylamine hydrobromide
≤0.5% tributylamine

mp

102-106 °C (lit.)

functional group

amine

greener alternative category

SMILES string

[Br-].CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.BrH/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;/h5-16H2,1-4H3;1H/q+1;/p-1

InChI key

JRMUNVKIHCOMHV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tetrabutylammonium bromide (TBAB) is a quaternary ammonium compound.[1] It is the most widely used phase transfer catalyst.[2] Its interfacial properties have been studied in case of hydroxide initiated reactions. This may be applied in understanding the mechanism of phase transfer reactions.[1] TBAB is reported to decrease retention time and remove peak tailing by acting as an ion pair reagent during the chromatographic analysis of quaternary ammonium compounds.[2] In the molten state TBAB behaves like an ionic liquid which is a promising green alternative to organic solvents in organic synthesis.[3][4] Its molar heat capacity, entropy and free energy function have been determined.[5]
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalysis. Click here for more information.

Application

Tetrabutylammonium bromide (TBAB) may be used in the molten state in the following processes:
  • Synthesis of (2S)-5-(3-phenyl-2-phthalimidylpropanoylamino)isophthalic acid.[6]
  • Synthesis of alkyl-substituted pyrroles in the absence of catalyst and organic solvent.[7]
  • Synthesis of dithioacetals from acetals by transthioacetalisation in a solvent free environment.[8]
  • Synthesis of polyamides (PAs) by the polymerization of terephthalic acid and diisocyanates.[4]
  • Catalyze the addition of thiols to conjugated alkenes.[9]
  • Dehydrochlorination of poly(vinyl chloride).[10]
Used with phosphorus pentoxide for greener deoxybromination.
Used with phosphorus pentoxide for greener deoxybromination.
Process for Producing Halogenated Heteroaryl Compounds

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chi Chen et al.
Nature communications, 13(1), 4935-4935 (2022-08-24)
Control over the copy number and nanoscale positioning of quantum dots (QDs) is critical to their application to functional nanomaterials design. However, the multiple non-specific binding sites intrinsic to the surface of QDs have prevented their fabrication into multi-QD assemblies
Heat capacities and thermodynamic properties of two tetramethylammonium halides.
Chang SS and Westrum Jr EF.
J. Chem. Phys. , 36(9), 2420-2423 (1962)
Silvia Ghimenti et al.
Scientific reports, 10(1), 7441-7441 (2020-05-06)
Heart failure (HF) is a cardiovascular disease affecting about 26 million people worldwide costing about $100 billons per year. HF activates several compensatory mechanisms and neurohormonal systems, so we hypothesized that the concomitant monitoring of a panel of potential biomarkers
Novel biobased polyurethanes synthesized from nontoxic phenolic diol containing l-tyrosine moiety under green media.
Mallakpour S, et al.
Journal of Polymers and the Environment, 18(4), 685-695 (2010)
Catalysis by ionic liquids: solvent-free efficient transthioacetalisation of acetals by molten tetrabutylammonium bromide.
Ranu BC, et al.
Journal of the Chemical Society. Perkin Transactions 1, 13, 1520-1522 (2002)

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Hi, what is the shelf life of this material?

    1 answer
    1. This product is not assigned an expiration date or recommended retest date. Products with no expiration date or recommended retest date should be routinely inspected by customers to ensure they perform as expected. These products are also subject to a one year warranty from the date of shipment. For more information you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/quality-services.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service