Skip to Content
Merck
All Photos(1)

Key Documents

349674

Sigma-Aldrich

Dimethyl cis-1,2,3,6-tetrahydrophthalate

99%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€153.00

€153.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€153.00

About This Item

Empirical Formula (Hill Notation):
C10H14O4
CAS Number:
Molecular Weight:
198.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€153.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.472 (lit.)

bp

141-142 °C/20 mmHg (lit.)

density

1.145 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

COC(=O)[C@H]1CC=CC[C@H]1C(=O)OC

InChI

1S/C10H14O4/c1-13-9(11)7-5-3-4-6-8(7)10(12)14-2/h3-4,7-8H,5-6H2,1-2H3/t7-,8+

InChI key

DVVAGRMJGUQHLI-OCAPTIKFSA-N

General description

Epoxidation of dimethyl cis-1,2,3,6-tetrahydrophthalate has been reported.[1] Desymmetrization of dimethyl cis-1,2,3,6-tetrahydrophthalate to (1S,2R)-1-methyl-cis-1,2,3,6-tetra-hydrophthalate has been reported.[2]

Application

Dimethyl cis-1,2,3,6-tetrahydrophthalate may be used in chemical synthesis.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereospecific epoxidation of dihydrophthalates.
Cerefice SA and Fields EK.
The Journal of Organic Chemistry, 41(2), 355-361 (1976)
The use of enzymes in organic synthesis and the life sciences: perspectives from the Swiss Industrial Biocatalysis Consortium (SIBC).
Meyer H-P, et al.
Catalysis Science & Technology, 3(1), 29-40 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service