Skip to Content
Merck
All Photos(1)

Documents

8.20370

Sigma-Aldrich

1,2,3,6-Tetrahydrophthalimide

for synthesis

Synonym(s):

1,2,3,6-Tetrahydrophthalimide, 4-Cyclohexene-1,2-dicarboxylic acid imide, 4-Cyclohexene-1,2-dicarboximide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9NO2
CAS Number:
Molecular Weight:
151.16
UNSPSC Code:
12352115
EC Index Number:
201-602-8
NACRES:
NA.22

Quality Level

Assay

≥98.0% (acidimetric)

form

solid

mp

133-137 °C

transition temp

flash point 178 °C

storage temp.

2-30°C

InChI

1S/C8H9NO2/c10-7-5-3-1-2-4-6(5)8(11)9-7/h1-2,5-6H,3-4H2,(H,9,10,11)

InChI key

CIFFBTOJCKSRJY-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 98.0 %
Melting range (lower value): ≥ 133 °C
Melting range (upper value): ≤ 137 °C
Identity (IR): passes test

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

370.0 °F - closed cup

Flash Point(C)

187.8 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service