Skip to Content
Merck
All Photos(1)

Key Documents

QBD10505

Sigma-Aldrich

Azido-dPEG®12-NHS ester

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€305.00
1000 MG
€1,030.00

€305.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€305.00
1000 MG
€1,030.00

About This Item

Empirical Formula (Hill Notation):
C31H56N4O16
Molecular Weight:
740.79
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

€305.00


Check Cart for Availability

Request a Bulk Order

Assay

>90%

form

solid or viscous liquid

reaction suitability

reaction type: click chemistry
reagent type: cross-linking reagent
reaction type: click chemistry

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(CCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCN=[N+]=[N-])ON1C(CCC1=O)=O

InChI key

JSNJRDFOCXNHTB-UHFFFAOYSA-N

Features and Benefits

The azido-dPEG®12-NHS ester contains an azide function on one end of a single molecular weight dPEG® spacer (47.2 Å) and NHS ester group on the other end of the spacer. The dPEG® spacer is hydrophilic and non-immunogenic and improves the water solubility of the target molecule while reducing the immunogenicity and increasing the hydrodynamic volume of the target. N-hydroxysuccinimide (NHS) ester readily reacts with amines in aqueous solution or organic solvent. The azide group reacts with an alkyne in the well-known click chemistry reaction. The click chemistry reaction proceeds by copper(I) or ruthenium catalysis or in a strain-catalyzed reaction with certain types of alkyne partners.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service