Skip to Content
Merck
All Photos(1)

Key Documents

QBD10119

Sigma-Aldrich

Acid-dPEG®9-NHS ester

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€325.00
1000 MG
€1,840.00

€325.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€325.00
1000 MG
€1,840.00

About This Item

Empirical Formula (Hill Notation):
C26H45NO15
Molecular Weight:
611.63
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

€325.00


Check Cart for Availability

Request a Bulk Order

Assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: cross-linking reagent
reactivity: amine reactive

functional group

NHS ester
carboxylic acid

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

OC(CCOCCOCCOCCOCCOCCOCCOCCOCCOCCC(ON1C(CCC1=O)=O)=O)=O

InChI

1S/C26H45NO15/c28-23-1-2-24(29)27(23)42-26(32)4-6-34-8-10-36-12-14-38-16-18-40-20-22-41-21-19-39-17-15-37-13-11-35-9-7-33-5-3-25(30)31/h1-22H2,(H,30,31)

InChI key

BJOKQEMLFQBOQZ-UHFFFAOYSA-N

Features and Benefits

Acid-dPEG®9-NHS ester is a unique heterobifunctional dPEG® crosslinking product which allows conjugation to two different moieties, each containing free amines. Because the single molecular weight dPEG® spacers contain an N-hydroxysuccinimide ester only on one end of the spacer, greater control of the conjugation reaction can be maintained. The free acid end of the spacer can be left unreacted to modify the surface properties of the molecule to which it is conjugated, or the free acid end can be activated and conjugated to a different group at a later time. The dPEG® spacers provide precise spacing/distance control, and the dPEG® itself provides the well-known benefits of increased water solubility; decreased immunogenicity; protection from proteolysis; increased hydrodynamic volume; and increased serum half-life.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service