Skip to Content
Merck
All Photos(1)

Documents

803332

Sigma-Aldrich

Sulfo-SANPAH (sulfosuccinimidyl 6-(4′-azido-2′-nitrophenylamino)hexanoate)

Synonym(s):

(sulfosuccinimidyl 6-(4′-azido-2′-nitrophenylamino)hexanoate), 1-[[6-[(4-azido-2-nitrophenyl)amino]-1-oxohexyl]oxy]-2,5-dioxo-3-pyrrolidinesulfonic acid monosodium salt, Sulfo-SANPAH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17O9N6NaS
Molecular Weight:
492.40
UNSPSC Code:
12352102
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

mol wt

492.4

reaction suitability

reagent type: cross-linking reagent

storage condition

desiccated

solubility

water: soluble

functional group

azide

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C1CC(S(=O)([O-])=O)C(N1OC(CCCCCNC2=CC=C(N=[N+]=[N-])C=C2[N+]([O-])=O)=O)=O.[Na+]

InChI

1S/C16H18N6O9S.Na/c17-20-19-10-5-6-11(12(8-10)22(26)27)18-7-3-1-2-4-15(24)31-21-14(23)9-13(16(21)25)32(28,29)30;/h5-6,8,13,18H,1-4,7,9H2,(H,28,29,30);/q;+1/p-1

InChI key

XXUXLXCHYVHAOD-UHFFFAOYSA-M

General description

ANB-NOS and Sulfo-SANPAH are heterobifunctional crosslinkers that contain an amine-reactive N-hydroxysuccinimide (NHS) ester and a photoactivatable nitrophenyl azide. NHS esters react efficiently with primary amino groups (-NH2) in pH 7-9 buffers to form stable amide bonds. When exposed to UV light nitrophenyl azides form a nitrene group that can initiate addition reactions with double bonds, insertion into C-H and N-H sites, or subsequent ring expansion to react with a nucleophile (e.g., primary amines).

Application


  • Dynamic Control of Cell Adhesion on a Stiffness-Tunable Substrate for Analyzing the Mechanobiology of Collective Cell Migration: This study uses Sulfo-SANPAH for photochemical cross-linking in cell adhesion research (Kamimura et al., 2017).

  • Evaluation of L929 fibroblast attachment and proliferation on ArgGlyAspSer (RGDS) immobilized chitosan: This paper investigates the use of Sulfo-SANPAH to enhance fibroblast attachment on chitosan substrates (Karakecili et al., 2016).

Features and Benefits

  • Reactive groups: sulfo-NHS ester and nitrophenyl azide
  • Reactive towards: amino groups and any nucleophile
  • Non-cleavable
  • N-Sulfosuccinimidyl-6-(4′-azido-2′-nitrophenylamino) hexanoate
  • Optimal photolysis occurs at 320-350 nm; minimizes damage to biomolecules by irradiation
  • Water soluble; non-cleavable

Caution

This product is sensitive to moisture. The vial is packaged in a resealable bag with a desiccant to reduce exposure to moisture. After cold storage, equilibrate the vial to room temperature before opening to reduce condensation inside the vial. Make fresh solutions. Storage of stock solutions is not recommended. After use, return the vial to the resealable bag. Close the bag and store the product at the recommended temperature.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christianne Gaudet et al.
Biophysical journal, 85(5), 3329-3335 (2003-10-29)
We examine the relationships of three variables (projected area, migration speed, and traction force) at various type I collagen surface densities in a population of fibroblasts. We observe that cell area is initially an increasing function of ligand density, but
F M Uckun et al.
Science (New York, N.Y.), 267(5199), 886-891 (1995-02-10)
B-cell precursor (BCP) leukemia is the most common form of childhood cancer and the second most common form of acute leukemia in adults. Human BCP leukemia was treated in a severe combined immunodeficient mouse model by targeting of the tyrosine
Xiao-Feng Zhang et al.
The Journal of cell biology, 218(7), 2329-2349 (2019-05-28)
Serotonin (5-HT) is known to increase the rate of growth cone advance via cofilin-dependent increases in retrograde actin network flow and nonmuscle myosin II activity. We report that myosin II activity is regulated by PKC during 5-HT responses and that
Yuehua Yang et al.
Biophysical journal, 119(7), 1427-1438 (2020-09-09)
Cell durotaxis is an essential process in tissue development. Although the role of cytoskeleton in cell durotaxis has been widely studied, whether cell volume and membrane tension are implicated in cell durotaxis remains unclear. By quantifying the volume distribution during
Dariusz Lachowski et al.
Scientific reports, 9(1), 7299-7299 (2019-05-16)
Liver fibrosis is characterised by a dense and highly cross-linked extracellular matrix (ECM) which promotes progression of diseases such as hepatocellular carcinoma. The fibrotic microenvironment is characterised by an increased stiffness, with rigidity associated with disease progression. External stiffness is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service