Skip to Content
Merck
All Photos(1)

Key Documents

JKA5088

Sigma-Aldrich

N3-PEG-NHS

average MN 5,000, cross-linking reagent click chemistry amine reactive

Synonym(s):

N3-PEG-NHS, Azide-PEG-NHS, N3-PEG-NHS, Polyethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€743.00

€743.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€743.00

About This Item

Linear Formula:
N3CH2CH2O(CH2CH2O)nCH2COOC4H4O2N
UNSPSC Code:
12162002
NACRES:
NA.23

€743.00


Check Cart for Availability

Request a Bulk Order

Product Name

N3-PEG5K-NHS, average Mn 5,000

form

solid

mol wt

average Mn 5,000

reaction suitability

reagent type: cross-linking reagent
reaction type: click chemistry
reactivity: amine reactive

polymer architecture

shape: linear
functionality: heterobifunctional

Looking for similar products? Visit Product Comparison Guide

General description

Azide PEG NHS Ester. The Azide group may be reduced to amine by hydrogenolysis; Click chemistry PEG reagent for reaction with alkynes.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service