Skip to Content
Merck
All Photos(1)

Key Documents

712531

Sigma-Aldrich

O-[(N-Succinimidyl)succinyl-aminoethyl]-O′-methylpolyethylene glycol

average Mn 750

Synonym(s):

Polyethylene glycol, O-Methyl-O′-[(N-succinimidyl)succinyl-aminoethyl]polyethylene glycol, O-Methyl-O′-[2-(succinylamino)-ethyl]polyethylene glycol N-succinimidyl ester, Ome-PEG-NHS

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€89.40

€89.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€89.40

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.22

€89.40


Check Cart for Availability

Request a Bulk Order

form

crystals

Quality Level

mol wt

average Mn 750

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Kanjiro Miyata (The University of Tokyo, Japan) provides insights on the rational design of polymeric materials for “smart” oligonucleotide delivery.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service