Skip to Content
Merck
All Photos(1)

Key Documents

QBD10067

Sigma-Aldrich

Amino-dPEG®6-acid

Synonym(s):

1-amino-3,6,9,12,15,18-Hexaoxaheneicosan-21-oic acid, 3-[(17-Amino-3,6,9,12,15-pentaoxaheptadec-1-yl)oxy]propanoic acid, Polyethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

1000 MG
€619.00

€619.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1000 MG
€619.00

About This Item

Empirical Formula (Hill Notation):
C15H31NO8
CAS Number:
Molecular Weight:
353.41
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

€619.00


Check Cart for Availability

Request a Bulk Order

Assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: cross-linking reagent
reactivity: carboxyl reactive

functional group

amine
carboxylic acid

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

NCCOCCOCCOCCOCCOCCOCCC(O)=O

InChI

1S/C15H31NO8/c16-2-4-20-6-8-22-10-12-24-14-13-23-11-9-21-7-5-19-3-1-15(17)18/h1-14,16H2,(H,17,18)

InChI key

PVRGRRPCHFTMMD-UHFFFAOYSA-N

Features and Benefits

Amino-dPEG®6-acid is a novel amino acid that can be manipulated, modified, and used like most other amino acids. The single molecular weight dPEG® spacer arms between the terminal amine and the terminal carboxylic acid are of exact length (25.1 Å), allowing for precise spatial control, and they are hydrophilic and non-immunogenic, imparting greater water solubility and hydrodynamic volume to target molecules.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service