Skip to Content
Merck
All Photos(4)

Key Documents

DWK292612607

Duran® Connection Cap System Gl 45 With Red Pbt Screw Cap

Synonym(s):

292612607

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€270.00

€270.00


Check Cart for Availability


Select a Size

Change View
1 EACH
€270.00

About This Item

UNSPSC Code:
41123300
NACRES:
NB.17

€270.00


Check Cart for Availability

material

red PBT screw cap

feature

number of ports 1 (Stainless steel)

packaging

pkg of 1 ea

manufacturer/tradename

DURAN® 292612607

General description

A robust and resistant hose connection system for DURAN® GL 45 laboratory and media bottles of all sizes. The bottle cap adapter simplifies the aseptic transfer of liquids within closed and sterile systems. The push-on tubing connectors on the inside and outside of the cap are suitable for connecting flexible tubing with an internal diameter of 8.0 mm. The freely rotating stainless steel middle piece allows the bottle to be opened without having to disconnect the tube connections. Connections and middle piece are made of stainless steel type 316L (1.4404/S31603). The silicone seal and the red GL 45 screw cap made of PBT ensure a liquid-tight seal. Wide temperature application range (up to 180 °C). Autoclavable (at 121 or 134 °C). With lot specific Retrace Code.

Application

small volumes of liquid need to be sampled or dispensed, such as a media supply for small bioreactors, in a perfusion circuit for cell culture, or oxygenation of very small samples

Features and Benefits

for 45 GL bottles Construction materials are certified as approved for food contact, and offer a high temperature resistance (up to 180 °C). The caps are fully autoclavable (121 °C / 15 minutes) and reusable

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service