Skip to Content
Merck
All Photos(2)

Key Documents

914916

Sigma-Aldrich

Biotin-SS-tyramide

≥95%

Synonym(s):

N-(2-((3-(4-Hydroxyphenethylamino)-3-oxopropyl)disulfanyl)ethyl)-5-(2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide, Cleavable biotin-phenol, Cleavable biotin-tyramide linker

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€387.00

€387.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€387.00

About This Item

Empirical Formula (Hill Notation):
C23H34N4O4S3
Molecular Weight:
526.74
UNSPSC Code:
12352200
NACRES:
NA.22

€387.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

powder

storage temp.

−20°C

Application

Biotin-SS-tyramide is a cleavable, biotinylated reagent for tyramide signal amplification used in many applications including immunohistochemistry, in situ hybridization, electron microscopy, ELISA, and others. It can be used together with both chromogenic and fluorescence visualization methods. The linker can also be used for synthesis of chemical biology tools for labeling target proteins in biological experiments and assays. It possesses dual functionality: enrichment via the biotin and an embedded disulfide bridge for cleavage under reducing conditions or by glutathione in the lysosome.

Automate your Biotin tagging with Synple Automated Synthesis Platform (SYNPLE-SC002)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service