Skip to Content
Merck
All Photos(1)

Key Documents

QBD10194

Sigma-Aldrich

NHS-S-S-dPEG®4-biotin (cleavable)

Synonym(s):

2,5-Dioxo-1-pyrrolidinyl 29-[(3aS,4S,6aR)-hexahydro-2-oxo-1H-thieno[3,4-d]imidazol-4-yl]-9,25-dioxo-12,15,18,21-tetraoxa-4,5-dithia-8,24-diazanonacosanoate, Polyethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€247.00
500 MG
€1,210.00

€247.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€247.00
500 MG
€1,210.00

About This Item

Empirical Formula (Hill Notation):
C30H49N5O11S3
CAS Number:
Molecular Weight:
751.93
UNSPSC Code:
51171641
PubChem Substance ID:
NACRES:
NA.22

€247.00


Check Cart for Availability

Request a Bulk Order

Assay

>90%

form

solid or viscous liquid

reaction suitability

reaction type: Biotinylations
reagent type: cross-linking reagent

functional group

disulfide

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(NCCSSCCC(ON1C(CCC1=O)=O)=O)CCOCCOCCOCCOCCNC(CCCCC2C3NC(NC3CS2)=O)=O

InChI

1S/C30H49N5O11S3/c36-24(4-2-1-3-23-29-22(21-47-23)33-30(41)34-29)31-9-12-43-14-16-45-18-17-44-15-13-42-11-7-25(37)32-10-20-49-48-19-8-28(40)46-35-26(38)5-6-27(35)39/h22-23,29H,1-21H2,(H,31,36)(H,32,37)(H2,33,34,41)

InChI key

QEVJWOMRMWJABZ-UHFFFAOYSA-N

Features and Benefits

NHS-S-S-dPEG®4-biotin is used extensively to label molecules that contain a free amine with biotin. This dPEG® spacer imparts a high degree of water solubility to the biotin label. For biotinylated peptides and proteins, the dPEG® spacer dramatically reduces aggregation and precipitation of the modified molecule and reduces non-specific binding of the molecule, both of which are common problems with molecules that have been biotinylated with the widely used NHS-LC-biotin. NHS-S-S-dPEG®4-biotin contains a disulfide group in the linear chain, allowing the biotin to be cleaved reductively from the labeled molecule.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service