Skip to Content
Merck
All Photos(1)

Documents

SML3484

Sigma-Aldrich

Biotin-XX-tyramide

≥95% (HPLC)

Synonym(s):

1H-Thieno[3,4-d]imidazole-4-pentanamide, hexahydro-N-[6-[[6-[[2-(4-hydroxyphenyl)ethyl]amino]-6-oxohexyl]amino]-6-oxohexyl]-2-oxo-, (3aS,4S,6aR)- (9CI, ACI), Biotin-Ahx-AhxX-phenol, Biotin-Ahx-AhxX-tyramide, Biotin-LC-LC-tyramide, Biotin-XX-Tyr; BxxP, Biotin-XX-phenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H47N5O5S
CAS Number:
Molecular Weight:
589.79
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to off-white

storage temp.

-10 to -25°C

Biochem/physiol Actions

Biotin-XX-tyramide (biotin-XX-phenol; BxxP) is a membrane-impermeant variant of the proximity labeling probe biotin-tyramide (biotin-phenol; BP) that corresponds to BP with a long and polar polyamide linker. Similary to BP, BxxP gives robust biotinylation with HRP and H2O2, but unlike BP, BxxP no longer enters cells, yielding selective labeling of cell surface, but not intracellular, proteins.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sam Kint et al.
Nucleic acids research, 49(8), e43-e43 (2021-01-30)
Characterization of the epigenetic status of individual cells remains a challenge. Current sequencing approaches have limited coverage, and it is difficult to assign an epigenetic status to the transcription state of individual gene alleles in the same cell. To address
Ken H Loh et al.
Cell, 166(5), 1295-1307 (2016-08-28)
Cellular compartments that cannot be biochemically isolated are challenging to characterize. Here we demonstrate the proteomic characterization of the synaptic clefts that exist at both excitatory and inhibitory synapses. Normal brain function relies on the careful balance of these opposing
Santosh Renuse et al.
Journal of the American Society for Mass Spectrometry, 31(2), 394-404 (2020-01-16)
The use of biotin or biotin-containing reagents is an essential component of many protein purification and labeling technologies. Owing to its small size and high affinity to the avidin family of proteins, biotin is a versatile molecular handle that permits
Jiefu Li et al.
Cell, 180(2), 373-386 (2020-01-21)
Molecular interactions at the cellular interface mediate organized assembly of single cells into tissues and, thus, govern the development and physiology of multicellular organisms. Here, we developed a cell-type-specific, spatiotemporally resolved approach to profile cell-surface proteomes in intact tissues. Quantitative
Sebastian Ols et al.
Cell reports, 30(12), 3964-3971 (2020-03-27)
Although intramuscular (i.m.) administration is the most commonly used route for licensed vaccines, subcutaneous (s.c.) delivery is being explored for several new vaccines under development. Here, we use rhesus macaques, physiologically relevant to humans, to identify the anatomical compartments and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service