Skip to Content
Merck
All Photos(2)

Key Documents

913863

Sigma-Aldrich

Biotin-AEEA-tyramide

Synonym(s):

N-(2-(2-(2-(4-Hydroxyphenethylamino)-2-oxoethoxy)ethoxy)ethyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide, N-[2-(4-hydroxyphenyl)ethyl]-2-(2-(2-biotinamidoethoxy)ethoxy)acetamid, Biotin-AEEA-phenol, Biotin-PEG-tyramide

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€370.00

€370.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€370.00

About This Item

Empirical Formula (Hill Notation):
C24H36N4O6S
Molecular Weight:
508.63
UNSPSC Code:
12352200
NACRES:
NA.22

€370.00


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

storage temp.

−20°C

Application

Biotin-AEEA-tyramide is a reagent for tyramide signal amplification used in many applications including immunohistochemistry, in situ hybridization, electron microscopy, ELISA, and others. It can be used together with both chromogenic and fluorescence visualization methods and can be added to any other standard IHC protocol, reducing the use of other reagents.

Automate your Biotin tagging with Synple Automated Synthesis Platform (SYNPLE-SC002)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service