Skip to Content
Merck
All Photos(3)

Key Documents

792357

Sigma-Aldrich

MorDalphos Pd G3

Synonym(s):

(2-(Di-1-adamantylphosphino)morpholinobenzene)[2-(2′-amino-1,1′-biphenyl)]palladium(II) methanesulfonate, Mesyl(2-(di-1-adamantylphosphino)morpholinobenzene)[2-(2′-amino-1,1′-biphenyl)]palladium(II),, MorDalphos-G3-palladacycle

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€98.30
500 MG
€120.00
2 G
€379.00
5 G
€882.00

€98.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€98.30
500 MG
€120.00
2 G
€379.00
5 G
€882.00

About This Item

Empirical Formula (Hill Notation):
C43H55N2O4PPdS
CAS Number:
Molecular Weight:
833.37
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€98.30


Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

feature

generation 3

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

mp

245-250 °C

functional group

phosphine

SMILES string

NC1=C(C=CC=C1)C2=C([Pd]OS(=O)(C)=O)C=CC=C2.P([C@]34C[C@@H](C[C@@H](C4)C5)C[C@@H]5C3)([C@@]6(C[C@@H](C7)C8)C[C@H]7C[C@H]8C6)C9=CC=CC=C9N%10CCOCC%10

InChI

1S/C30H42NOP.C12H10N.CH4O3S.Pd/c1-2-4-28(27(3-1)31-5-7-32-8-6-31)33(29-15-21-9-22(16-29)11-23(10-21)17-29)30-18-24-12-25(19-30)14-26(13-24)20-30;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-5(2,3)4;/h1-4,21-26H,5-20H2;1-6,8-9H,13H2;1H3,(H,2,3,4);/q;;;+1/p-1

InChI key

XDLJAOATDSJDKP-UHFFFAOYSA-M

General description

MorDalphos Pd G3 is a third generation (G3) Buchwald precatalyst. It is air, moisture and thermally-stable and is highly soluble in a wide range of common organic solvents. It has long life in solutions. MorDalphos Pd G3 is an excellent reagent for palladium catalyzed cross-coupling reactions. Some of its unique features include lower catalyst loadings, shorter reaction time, efficient formation of the active catalytic species and accurate control of ligand: palladium ratio.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

All of the preformed catalysts used in the kit are air and moisture stable complexes in their commercially available form. Once activated by base under the reaction conditions they become sensitive to air. To best enable scale-up success, the use of standard Schlenk technique is recommended.

All contents in the foil bag are weighed, plated, packed, and sealed in a glove box under nitrogen.

G3 and G4 Buchwald palladium precatalysts are the newest air, moisture, and thermally stable crossing-coupling complexes used in bond formation for their versatility and high reactivity.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service