Skip to Content
Merck
All Photos(2)

Key Documents

804983

Sigma-Aldrich

DPPF Pd G3

95%

Synonym(s):

Methanesulfonato 1,1-ferrocenediyl-bis(diphenylphosphino) (2′-amino-1,1′-biphenyl-2-yl) palladium(II)

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€124.00
1 G
€366.00

€124.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€124.00
1 G
€366.00

About This Item

Empirical Formula (Hill Notation):
C47H41FeNO3P2PdS
CAS Number:
Molecular Weight:
924.11
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€124.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

solid

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

134-208 °C (MP data is all decomp.)

SMILES string

NC1=C(C2=C([Pd]OS(C)(=O)=O)C=CC=C2)C=CC=C1.P(C3=CC=CC=C3)([C]4[C][C][C][C]4)C5=CC=CC=C5.P(C6=CC=CC=C6)([C]7[C][C][C][C]7)C8=CC=CC=C8.[Fe]

InChI

1S/2C17H14P.C12H10N.CH4O3S.Fe.Pd/c2*1-3-9-15(10-4-1)18(17-13-7-8-14-17)16-11-5-2-6-12-16;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-5(2,3)4;;/h2*1-14H;1-6,8-9H,13H2;1H3,(H,2,3,4);;/q;;;;;+1/p-1

InChI key

ASVGFZMDZZREFR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Pd precatalyst for cross-coupling

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

G3 and G4 Buchwald palladium precatalysts are the newest air, moisture, and thermally stable crossing-coupling complexes used in bond formation for their versatility and high reactivity.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service