Skip to Content
Merck
All Photos(3)

Key Documents

903027

Sigma-Aldrich

QPhos Pd G3

Synonym(s):

QPhos G3 Palladacycle, QPhos Palladacycle, [2′-(Amino-κN)[1,1′-biphenyl]-2-yl-κC][1′-[bis(1,1-dimethylethyl)phosphino]-1,2,3,4,5-pentaphenylferrocene](methanesulfonato-κO)palladium

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€201.00
1 G
€566.00

€201.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€201.00
1 G
€566.00

About This Item

Empirical Formula (Hill Notation):
C61H60FeNO3PPdS
CAS Number:
Molecular Weight:
1080.44
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

€201.00


Check Cart for Availability

Request a Bulk Order

form

powder

feature

generation 3

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

functional group

phosphine

SMILES string

O=S(=O)([O-][Pd+2]1([C-]=2C=CC=CC2C=3C=CC=CC3[NH2]1)[P](C(C)(C)C)(C(C)(C)C)[C-]45[CH]6=[CH]7[CH]8=[CH]4[Fe+2]7869%10%11%125C=%13(C=%14C=CC=CC%14)C%12(C=%15C=CC=CC%15)=C%11(C=%16C=CC=CC%16)[C-]%10(C=%17C=CC=CC%17)C%139C=%18C=CC=CC%18)C

InChI

InChI=1S/C35H25.C13H22P.C12H10N.CH4O3S.Fe.Pd/c1-6-16-26(17-7-1)31-32(27-18-8-2-9-19-27)34(29-22-12-4-13-23-29)35(30-24-14-5-15-25-30)33(31)28-20-10-3-11-21-28;1-12(2,3)14(13(4,5)6)11-9-7-8-10-11;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-5(2,3)4;;/h1-25H;7-10H,1-6H3;1-6,8-9H,13H2;1H3,(H,2,3,4);;/q3*-1;;2*+2/p-1

InChI key

NNVAEYGNFLUPRU-UHFFFAOYSA-M

General description

QPhos Pd G3 is a third generation (G3) Buchwald precatalyst. It is air, moisture and thermally-stable and is highly soluble in a wide range of common organic solvents. It has long life in solutions. QPhos Pd G3 is an excellent reagent for palladium catalyzed cross-coupling reactions. Some of its unique features include lower catalyst loadings, shorter reaction time, efficient formation of the active catalytic species and accurate control of ligand: palladium ratio.

Application

QPhos Pd G3 has been used as a precatalyst for the arylboration of isoprene with regioselectivity. It may also be used for a variety of aminination and etherifications of aryl halides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin B Smith et al.
Journal of the American Chemical Society, 139(23), 7721-7724 (2017-06-06)
A method for the regioselective arylboration of isoprene and its derivatives is presented. These reactions allow for the synthesis of useful building blocks from simple components. Through these studies, an unusual additive effect with DMAP has been uncovered that allows
Thomas J Greshock et al.
Angewandte Chemie (International ed. in English), 55(44), 13714-13718 (2016-10-22)
The reactivity of a representative set of 17 organozinc pivalates with 18 polyfunctional druglike electrophiles (informers) in Negishi cross-coupling reactions was evaluated by high-throughput experimentation protocols. The high-fidelity scaleup of successful reactions in parallel enabled the isolation of sufficient material

Articles

G3 and G4 Buchwald palladium precatalysts are the newest air, moisture, and thermally stable crossing-coupling complexes used in bond formation for their versatility and high reactivity.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service