Skip to Content
Merck
All Photos(2)

Documents

900592

Sigma-Aldrich

(PAd-DalPhos)NiCl(otol)

Synonym(s):

[8-[2-[bis(2-methylphenyl)phosphino-κP]phenyl]-1,3,5,7-tetramethyl-2,4,6-trioxa-8-phosphatricyclo[3.3.1.13,7]decane-κP8]chloro(2-methylphenyl)-, (SP-4-2)-Nickel

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H41ClNiO3P2
CAS Number:
Molecular Weight:
689.81
UNSPSC Code:
12352200
NACRES:
NA.22

form

powder

Quality Level

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

SMILES string

CC(C=CC=C1)=C1P2(C3=C(C)C=CC=C3)C4=CC=CC=C4P5([Ni]2(C6=C(C)C=CC=C6)Cl)[C@](C)(O7)C[C@]8(C)O[C@@]7(C)C[C@](C)5O8

Application

Air stable Ni catalyst for C(sp2)-N coupling.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service