Skip to Content
Merck
All Photos(1)

Documents

544086

Sigma-Aldrich

3,3,3-Trifluoro-1-propyne

97%

Synonym(s):

1,1,1-Trifluoropropyne, TFP, Trifluoromethylethyne

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C≡CH
CAS Number:
Molecular Weight:
94.04
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

SMILES string

FC(F)(F)C#C

InChI

1S/C3HF3/c1-2-3(4,5)6/h1H

InChI key

PRDFNJUWGIQQBW-UHFFFAOYSA-N

Packaging

Supplied in a carbon steel lecture bottle with a CGA180M/CGA110F needle valve installed.

Compatible with the following:
  • Aldrich® lecture-bottle station systems
  • Aldrich® lecture-bottle gas regulators

Other Notes

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

hose barb

Product No.
Description
Pricing

purge valve

Product No.
Description
Pricing

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service