Key Documents
Z562599
Sigma-Aldrich
Aldrich® lecture-bottle station
for non-corrosive gases, brass regulator
Synonym(s):
Lecture Bottle Station
Select a Size
1 EA
€1,510.00
€1,510.00
Estimated to ship on28 March 2025
Select a Size
Change View
1 EA
€1,510.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
€1,510.00
Estimated to ship on28 March 2025
material
brass regulator
inlet CGA
180
General description
Dispensing lecture-bottle gases has never been safer or simpler using these portable lecture-bottle stations. The premium quality components housed within a station are matched to the application, fully assembled, leak-tested, and ready for use.
Just connect the station to the lecture-bottle, process, and a source of inert gas, if using a station with a purge valve, and you are ready to go.
Safe and Simple:
  • Gas dispensing and storage in one compact, portable system
  • Keeps lecture-bottles upright and stable, eliminates tip-over
  • Reduces equipment damage and chance of personal injury
  • Choice of four models matched to the application
  • Online Lecture-Bottle Equipment to Gases Selection Guide
Station Specifications:
1. Heavy-gauge steel cabinet with durable, red enamel finish, table-top or wall mountable
2. Quality, panel-mounted regulators and purge valves
3. Braided, 316L stainless steel hose with CGA-180 remote connection to the lecture-bottle
4. Support for two lecture-bottles, one in use and a spare
5. Clearly labeled gauges, gas connections, and valve functions
6. Instructions for Use label on front of cabinet
7. Fully assembled, leak-tested and ready for use
For inert and non-corrosive gas applications.
Regulator specifications:
Single stage, panel-mounted brass body with stainless steel pressure diaphragm, brass nozzle, polyurethane seat, nylon seals, self seating type relief valve, 11/2 in. diameter gauges, 3000 psig maximum inlet pressure, 0-80 psig delivery range, brass outlet needle valve with 1/4 in. NPTF connection.
Linkage
For more information on station set-up, see the Lecture Bottle Technical Bulletin
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
recommended
Product No.
Description
Pricing
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)