Skip to Content
Merck
All Photos(5)

Key Documents

206032

Sigma-Aldrich

Platinum(IV) oxide

greener alternative

Synonym(s):

Adam’s catalyst, NSC 402624, Platinic oxide, Platinum dioxide

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€84.70
1 G
€214.00
5 G
€661.00
50 G
€6,110.00

€84.70


Estimated to ship on01 April 2025


Request a Bulk Order

Select a Size

Change View
250 MG
€84.70
1 G
€214.00
5 G
€661.00
50 G
€6,110.00

About This Item

Linear Formula:
PtO2
CAS Number:
Molecular Weight:
227.08
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€84.70


Estimated to ship on01 April 2025


Request a Bulk Order

form

solid

Quality Level

composition

Pt, 81-83%

reaction suitability

core: platinum
reagent type: catalyst

greener alternative product characteristics

Designing Safer Chemicals
Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

surface area

≥60 m2/g

mp

450 °C (lit.)

greener alternative category

SMILES string

O=[Pt]=O

InChI

1S/2O.Pt

InChI key

YKIOKAURTKXMSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Catalyst for hydrogenation of various functional groups with minimal hydrogenolysis problems; used as dehydrogenation catalyst; used for selective oxidation of primary alcohols.
Catalyst for:
  • Hydrohydrazination reactions
  • Oxidation of sugars
  • Low-temperature oxidation of carbon monoxide
  • Oxygen reduction reaction
  • Hydrogenation reactions
  • Hydrogenolysis of the cyclopropyl group into an isopropyl group
  • Hydroxycyclopropanation reactions
Platinum(IV) oxide has been used for the reduction of citernyl bromide to form (S)-3,7-dimethyloctylbromide.[1]

For small scale and high throughput uses, product is also available as ChemBeads (928291)

Pictograms

Flame over circle

Signal Word

Danger

Hazard Statements

Hazard Classifications

Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Just, G.; Wang, Z. Y.; Chan, L.
The Journal of Organic Chemistry, 53, 1030-1030 (1988)
Avery, M. A.; Verlander, M. S.; Goodman, M.
The Journal of Organic Chemistry, 45, 2750-2750 (1980)
Room-temperature discotic cholesteric and nematic phases: influence of 3, 7-dimethyloctane peripheral chain on the molecular self-assembly of radial polyalkynylbenzene.
Varshney SK
Liq. Cryst., 38(1), 53-60 (2011)
Lee, T. B. K.; Wong, G. S. K.
The Journal of Organic Chemistry, 56, 872-872 (1991)
Development of GSKDevelopment of GSK's reagent guides ? embedding sustainability into reagent selections reagent guides ? embedding sustainability into reagent selection.
Adams JP, et al.
Green Chemistry, 15, 1542-1549 (2013)

Articles

Oxidation and reduction reactions are some of the most common transformations encountered in organic synthesis

Questions

1–6 of 6 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. Is Product 206032, Platinum (IV) oxide, water soluble?

    1 answer
    1. It is insoluble in water.

      Helpful?

  3. Are there any safety concerns with Product 206032, Platinum (IV) oxide?

    1 answer
    1. After exposure to hydrogen, the resulting platinum black can be pyrophoric.  Therefore, it should not be allowed to dry and all exposure to oxygen should be minimized.

      Helpful?

  4. How can Product 206032, Platinum (IV) oxide, be used as a catalyst?

    1 answer
    1. Platinum(IV) oxide can be used as a catalyst in hydrogenations.  The actual catalyst is platinum black which is formed in situ by reduction of the PtO2 by the hydrogen used for the hydrogenation.  It is especially useful for reduction at room temperature and hydrogen pressures up to 4 atmospheres.  It is suitable for the reduction of double and triple bonds, aromatic nuclei, carbonyl groups, nitro groups, and nitriles.

      Helpful?

  5. In which solvents is Product 206032, Platinum (IV) oxide, soluble?

    1 answer
    1. Per the the chemicals encyclopedia published by the Royal Society of Chemistry, 13th Edition, when freshly precipitated, Platinum(IV) Oxide is soluble in concentrated acids and dilute phosphoric acid, especially when warmed.  It is easily soluble in dilute solutions of potassium hydroxide.

      Helpful?

  6. Can the Platinum be recovered from Product 206032, Platinum (IV) oxide?

    1 answer
    1. Internet sources do indicate that Platinum can be recovered from spent catalyst by conversion to ammonium chloroplatinate using aqua regia followed by ammonia.  However, we do have a process for doing so.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service