Skip to Content
Merck
All Photos(1)

Key Documents

412627

Sigma-Aldrich

5-tert-Butyl-o-anisidine

97%

Synonym(s):

5-tert-Butyl-2-methoxyaniline

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€54.10

€54.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€54.10

About This Item

Linear Formula:
(CH3)3CC6H3(OCH3)NH2
CAS Number:
Molecular Weight:
179.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€54.10


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

bp

204-208 °C (lit.)

mp

29-30 °C (lit.)

density

0.988 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(cc1N)C(C)(C)C

InChI

1S/C11H17NO/c1-11(2,3)8-5-6-10(13-4)9(12)7-8/h5-7H,12H2,1-4H3

InChI key

DLTLFDQLMFMTRQ-UHFFFAOYSA-N

Related Categories

Application

5-tert-Butyl-o-anisidine may be used for the preparation of anisole containing polypyridylamine, a potential tetradentate ligand.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heather R Lucas et al.
Journal of the American Chemical Society, 131(9), 3230-3245 (2009-02-17)
With an anisole-containing polypyridylamine potential tetradentate ligand (O)L, a mu-1,2-peroxo-dicopper(II) complex [{(O)LCu(II)}(2)(O(2)(2-))](2+) forms from the reaction of the mononuclear compound [Cu(I)((O)L)(MeCN)]B(C(6)F(5))(4) ((O)LCu(I)) with O(2) in noncoordinating solvents at -80 degrees C. Thermal decay of this peroxo complex in the presence

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service