Skip to Content
Merck
All Photos(1)

Key Documents

565830

Sigma-Aldrich

4-tert-Butylanisole

97%

Synonym(s):

4-Methoxy-tert-butylbenzene

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€65.20

€65.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€65.20

About This Item

Linear Formula:
(CH3)3CC6H4OCH3
CAS Number:
Molecular Weight:
164.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€65.20


Check Cart for Availability

Request a Bulk Order

Assay

97%

refractive index

n20/D 1.503 (lit.)

density

0.938 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(cc1)C(C)(C)C

InChI

1S/C11H16O/c1-11(2,3)9-5-7-10(12-4)8-6-9/h5-8H,1-4H3

InChI key

MCUPBIBNSTXCPQ-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qingqing Mei et al.
Science advances, 4(5), eaaq0266-eaaq0266 (2018-05-26)
Ether bond activation is very interesting because the synthesis of many valuable compounds involves conversion of ethers. Moreover, C-O bond cleavage is also very important for the transformation of biomass, especially lignin, which abundantly contains ether bonds. Developing efficient methods

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service