Skip to Content
Merck
All Photos(1)

Key Documents

M29355

Sigma-Aldrich

2-Methylanisole

99%

Synonym(s):

2-Methyl-1-methoxybenzene, o-Cresol methyl ether, o-Cresyl methyl ether, o-Methoxytoluene, o-Methylanisole

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€141.00

€141.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€141.00

About This Item

Linear Formula:
CH3C6H4OCH3
CAS Number:
Molecular Weight:
122.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€141.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

refractive index

n20/D 1.516 (lit.)

bp

170-172 °C (lit.)

density

0.985 g/mL at 25 °C (lit.)

SMILES string

COc1ccccc1C

InChI

1S/C8H10O/c1-7-5-3-4-6-8(7)9-2/h3-6H,1-2H3

InChI key

DTFKRVXLBCAIOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elodie Guyonnet Bilé et al.
ChemSusChem, 5(1), 91-101 (2012-01-18)
Optically active amphiphilic compounds derived from N-methylephedrine, N-methylprolinol, or cinchona derivatives possessing bromide or chiral lactate counterions were efficiently used as protective agents for rhodium(0) nanoparticles. The full characterization of these surfactants and the obtained nanocatalysts was performed by means
L Higgins et al.
Archives of biochemistry and biophysics, 385(1), 220-230 (2001-05-22)
Regioselectivity is used to determine the absolute energetic differences for four different reactions catalyzed by P450. Abstraction of a hydrogen from a benzylic carbon containing a chlorine has a 1.0 kcal/mol lower barrier than abstraction from a simple benzylic carbon
Graziela G Bianco et al.
The Journal of organic chemistry, 74(6), 2561-2566 (2009-02-24)
The first synthesis of the natural product (+)-mutisianthol was accomplished in 11 steps and in 21% overall yield from 2-methylanisole. The synthesis of its enantiomer was also performed in a similar overall yield. The absolute configuration of the sesquiterpene (+)-mutisianthol
Johannes Niebler et al.
Phytochemistry, 109, 66-75 (2014-12-04)
Frankincense has been known, traded and used throughout the ages for its exceptional aroma properties, and is still commonly used in both secular and religious settings to convey a pleasant odor. Surprisingly, the odoriferous principle(s) underlying its unique odor profile

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service