Skip to Content
Merck
All Photos(2)

Key Documents

247103

Sigma-Aldrich

2-Bromo-5-nitrotoluene

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€63.30

€63.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€63.30

About This Item

Linear Formula:
CH3C6H3(NO2)Br
CAS Number:
Molecular Weight:
216.03
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€63.30


Check Cart for Availability

Request a Bulk Order

Assay

98%

mp

78-80 °C (lit.)

functional group

bromo
nitro

SMILES string

Cc1cc(ccc1Br)[N+]([O-])=O

InChI

1S/C7H6BrNO2/c1-5-4-6(9(10)11)2-3-7(5)8/h2-4H,1H3

InChI key

HIMGPQVBNICCGL-UHFFFAOYSA-N

Related Categories

Application

2-Bromo-5-nitrotoluene has been used in the synthesis of :
  • 4′-amino-4-hydroxy-2′-methylbiphenyl[1]
  • 3′:4-dinitro-2-methyldiphenyl ether via condensation with potassium m-nitrophenoxide[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

16. Substitution in the methyl-4'-nitro-and-4'-acetamido-diphenyl ethers.
Scarborough HA and Sweeten JL.
Journal of the Chemical Society, 52-56 (1934)
Dimorphs of 4'-amino-4-hydroxy-2'-methylbiphenyl: Assessment of likelihood of polymorphism in flexible molecules.
Dey A and Desiraju GR.
CrystEngComm, 8(6), 477-481 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service