Skip to Content
Merck
All Photos(2)

Key Documents

294756

Sigma-Aldrich

Iodomethane-13C,d3

99.5 atom % D, 99 atom % 13C, 99% (CP), contains copper as stabilizer

Synonym(s):

13C and D Labeled iodomethane, Methyl-13C,d3 iodide

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€285.00
5 G
€969.00

€285.00


Estimated to ship on12 April 2025


Request more information

Select a Size

Change View
1 G
€285.00
5 G
€969.00

About This Item

Linear Formula:
13CD3I
CAS Number:
Molecular Weight:
145.95
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.12

€285.00


Estimated to ship on12 April 2025


Request more information

vapor density

4.89 (vs air)

Quality Level

vapor pressure

332 mmHg ( 20 °C)

isotopic purity

99 atom % 13C
99.5 atom % D

Assay

99% (CP)

form

liquid

contains

copper as stabilizer

technique(s)

protein expression: suitable

bp

42 °C (lit.)

mp

-66.5 °C (lit.)

density

2.340 g/mL at 25 °C

application(s)

environmental

mass shift

M+4

storage temp.

2-8°C

SMILES string

[2H][13C]([2H])([2H])I

InChI

1S/CH3I/c1-2/h1H3/i1+1D3

InChI key

INQOMBQAUSQDDS-KQORAOOSSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Carc. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service