Skip to Content
Merck
All Photos(1)

Key Documents

372080

Sigma-Aldrich

Methyl-d3-lithium, as complex with lithium iodide solution

0.5 M in diethyl ether, 99 atom % D

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
€181.00

€181.00


Check Cart for Availability

Request more information

Select a Size

Change View
50 ML
€181.00

About This Item

Linear Formula:
CD3Li · LiI
Molecular Weight:
158.84
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

€181.00


Check Cart for Availability

Request more information

isotopic purity

99 atom % D

Quality Level

form

liquid

concentration

0.5 M in diethyl ether

density

0.816 g/mL at 25 °C

mass shift

M+3

storage temp.

2-8°C

SMILES string

[Li+].[I-].[2H]C([2H])([2H])[Li]

InChI

1S/CH3.HI.2Li/h1H3;1H;;/q;;;+1/p-1/i1D3;;;

InChI key

LHJHNKBQWVOCIH-AGUGZIQGSA-M

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service