Skip to Content
Merck
All Photos(2)

Key Documents

176001

Sigma-Aldrich

Methyl-d3-amine hydrochloride

99 atom % D

Synonym(s):

Deuterated methylamine hydrochloride, Methyl-d3-ammonium chloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€165.00

€165.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 G
€165.00

About This Item

Linear Formula:
CD3NH2·HCl
CAS Number:
Molecular Weight:
70.54
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

€165.00


Check Cart for Availability

Request more information

isotopic purity

99 atom % D

Quality Level

form

powder

mp

232-234 °C (lit.)

application(s)

pharmaceutical

mass shift

M+3

SMILES string

Cl.[2H]C([2H])([2H])N

InChI

1S/CH5N.ClH/c1-2;/h2H2,1H3;1H/i1D3;

InChI key

NQMRYBIKMRVZLB-NIIDSAIPSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Der-Yen Lee et al.
Journal of chromatography. A, 1388, 60-68 (2015-03-04)
Electrospray ionization (ESI) is the most useful interface for mass spectrometry associated with liquid chromatography (LC). However, analyses of polar metabolites become a challenge because the high polarity impairs the separation of metabolites in reversed-phase liquid chromatography and ionization of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service