The z693847 product is the same as the Bel-Art F38952-0000 product. According to Bel-Art's website page, this ampoule breaker can open 1ml, 2ml, 5ml, and 10 ml ampules. Link: https://www.belart.com/bel-art-f38952-0000-break-safe-ampule-opener-3-x-3-x-3.html
Z693847
Scienceware® Break-Safe™ ampule opener
allows one handed opening of up to 3 ampules at a time
Synonym(s):
ampule breakers, glass ampoule openers
Select a Size
€70.50
Estimated to ship on01 April 2025
Select a Size
About This Item
€70.50
Estimated to ship on01 April 2025
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F38952-0000
L × W × H
9.5 cm (3.75 in.) × 7.6 cm (3 in.) × 8.3 cm (3.25 in.)
General description
After removing ampule contents, turn the device upside down over a trash receptacle to safely discard ampules and tops.
- Includes 4 inserts to allow opening of 1 mL, 2 mL, 5 mL or 10 mL ampules
- Acrylic copolymer plastic construction has broad chemical resistance
- Transparency allows unobstructed viewing of ampule contents at all times
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
Already Own This Product?
Customers Also Viewed
Protocols
Scienceware® Break-Safe™ ampule opener
-
Dear madam, dear sir, I have a question about (Scienceware® Break-Safe™ ampule opener), Can this product open 10 ml and 20 ml ampoules Waiting your answer
1 answer-
Helpful?
-