Skip to Content
Merck
All Photos(1)

Key Documents

287407

Sigma-Aldrich

Vitamin K1

98%

Synonym(s):

2-Methyl-3-phytyl-1,4-naphthoquinone, 3-Phytylmenadione, Phylloquinone, Vitamin K1(20)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H46O2
CAS Number:
Molecular Weight:
450.70
Beilstein:
2568816
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

98%

refractive index

n20/D 1.527 (lit.)

mp

−20 °C (lit.)

density

0.984 g/mL at 25 °C (lit.)

SMILES string

O=C(C1=CC=CC=C21)C(C)=C(C/C=C(CCC[C@@H](CCC[C@H](C)CCCC(C)C)C)\C)C2=O

InChI

1S/C31H46O2/c1-22(2)12-9-13-23(3)14-10-15-24(4)16-11-17-25(5)20-21-27-26(6)30(32)28-18-7-8-19-29(28)31(27)33/h7-8,18-20,22-24H,9-17,21H2,1-6H3/b25-20+/t23-,24-/m1/s1

InChI key

MBWXNTAXLNYFJB-NKFFZRIASA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Yoshitomo Suhara et al.
Journal of medicinal chemistry, 54(13), 4918-4922 (2011-05-31)
Vitamin K(2) has been demonstrated to induce gene expression related to bone formation through a nuclear steroid and xenobiotic receptor (SXR). We synthesized new vitamin K analogues with the same isoprene side chains symmetrically introduced at the 2 and 3
Sanjeev Kumar et al.
Journal of medicinal chemistry, 51(6), 1706-1718 (2008-03-06)
Indoleamine 2,3-dioxygenase (IDO) is emerging as an important new therapeutic target for the treatment of cancer, chronic viral infections, and other diseases characterized by pathological immune suppression. While small molecule inhibitors of IDO exist, there remains a dearth of high-potency
A A Toropov et al.
European journal of medicinal chemistry, 43(4), 714-740 (2007-07-17)
Simplified molecular input line entry system (SMILES) has been utilized in constructing quantitative structure-property relationships (QSPR) for octanol/water partition coefficient of vitamins and organic compounds of different classes by optimal descriptors. Statistical characteristics of the best model (vitamins) are the
Zhichao Liu et al.
PLoS computational biology, 7(12), e1002310-e1002310 (2011-12-24)
Drug-induced liver injury (DILI) is a significant concern in drug development due to the poor concordance between preclinical and clinical findings of liver toxicity. We hypothesized that the DILI types (hepatotoxic side effects) seen in the clinic can be translated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service