Merck
  • Indoleamine 2,3-dioxygenase is the anticancer target for a novel series of potent naphthoquinone-based inhibitors.
Indoleamine 2,3-dioxygenase is the anticancer target for a novel series of potent naphthoquinone-based inhibitors.
Journal of medicinal chemistry (2008-03-06)
Sanjeev Kumar, William P Malachowski, James B DuHadaway, Judith M LaLonde, Patrick J Carroll, Daniel Jaller, Richard Metz, George C Prendergast, Alexander J Muller
ABSTRACT
Indoleamine 2,3-dioxygenase (IDO) is emerging as an important new therapeutic target for the treatment of cancer, chronic viral infections, and other diseases characterized by pathological immune suppression. While small molecule inhibitors of IDO exist, there remains a dearth of high-potency compounds offering in vivo efficacy and clinical translational potential. In this study, we address this gap by defining a new class of naphthoquinone-based IDO inhibitors exemplified by the natural product menadione, which is shown in mouse tumor models to have similar antitumor activity to previously characterized IDO inhibitors. Genetic validation that IDO is the critical in vivo target is demonstrated using IDO-null mice. Elaboration of menadione to a pyranonaphthoquinone has yielded low nanomolar potency inhibitors, including new compounds which are the most potent reported to date (K(i) = 61-70 nM). Synthetic accessibility of this class will facilitate preclinical chemical-genetic studies as well as further optimization of pharmacological parameters for clinical translation.
Product Description
Supelco
Phylloquinone (K1), analytical standard
Supelco
p-Benzoquinone, for spectrophotometric det. of amines, ≥99.5% (HPLC)
Sigma-Aldrich
p-Benzoquinone, reagent grade, ≥98%
Supelco
Menadione (K3), analytical standard
Sigma-Aldrich
Methyl-p-benzoquinone, 98%
Sigma-Aldrich
Chromone, 99%
Sigma-Aldrich
1,4-Naphthoquinone, purum, ≥96.5% (HPLC)
Sigma-Aldrich
5-Hydroxy-1,4-naphthoquinone, 97%
Sigma-Aldrich
1,4-Naphthoquinone, 97%
Sigma-Aldrich
Vitamin K1, BioXtra, ≥99.0% (sum of isomers, HPLC), mixture of isomers
Sigma-Aldrich
Menadione, crystalline
Sigma-Aldrich
Vitamin K1, viscous liquid
Sigma-Aldrich
Menadione, meets USP testing specifications